Learn More
Aviva Systems Biology Corp GLYATL2 Antibody - middle region (ARP49019_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49019_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLYATL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GLYATL2. Alternate names: BXMAS2-10, GATF-B, MGC24009ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59995