Learn More
MSC Betco Disinfectant, Aerosol, Citrus Bouquet, Tuberculocidal, 15.5 oz
Catalog No. 502839225
Encompass
This economical, ready-to-use aerosol disinfectant provides powerful disinfecting and deodorizing performance. It is designed for daily maintenance in various settings including restrooms, hospitals, households, and garbage cans. This tuberculocidal disinfectant effectively cleans multiple hard, non-porous surfaces.
  • Economical, ready-to-use aerosol
  • Powerful disinfecting and deodorizing performance
  • Tuberculocidal disinfectant for hard, non-porous surfaces
  • Kills a wide range of viruses and bacteria including Influenza A, B, Salmonella, MRSA, VRE, and HIV
  • CFC-free formula
  • Prevents mold and mildew
  • Meets AOAC efficacy standards for hospital aerosol disinfectants
  • Sanitizes by killing 99.9% of Staphylococcus aureus and Enterobacter aerogenes
Catalog No. 50-283-9225 Supplier MSC Supplier No. 72969272
This economical, ready-to-use aerosol disinfectant provides powerful disinfecting and deodorizing performance. It is designed for daily maintenance in various settings including restrooms, hospitals, households, and garbage cans. This tuberculocidal disinfectant effectively cleans multiple hard, non-porous surfaces.
  • Economical, ready-to-use aerosol
  • Powerful disinfecting and deodorizing performance
  • Tuberculocidal disinfectant for hard, non-porous surfaces
  • Kills a wide range of viruses and bacteria including Influenza A, B, Salmonella, MRSA, VRE, and HIV
  • CFC-free formula
  • Prevents mold and mildew
  • Meets AOAC efficacy standards for hospital aerosol disinfectants
  • Sanitizes by killing 99.9% of Staphylococcus aureus and Enterobacter aerogenes