Learn More
Medchemexpress LLC Glyburide-d11 | 1189985-02-1 | C23H17D11ClN3O5S | 5 MG
SDP
Catalog No. 5000373664
Encompass_Preferred
Glyburide-d11 is the deuterium labeled Glibenclamide, an orally active ATP-sensitive K+ channel (KATP) inhibitor used in diabetes and obesity research. It inhibits P-glycoprotein and directly binds and blocks SUR1 subunits of KATP. This compound interferes with mitochondrial bioenergetics and can induce autophagy. It is suitable as a tracer and internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits P-glycoprotein
  • Binds and blocks SUR1 subunits of KATP
  • Interferes with mitochondrial bioenergetics
  • Can induce autophagy
Catalog No. 50-003-73664 Supplier Medchemexpress LLC Supplier No. HY15206S5MG
Add to Cart
Add to Cart
Glyburide-d11 is the deuterium labeled Glibenclamide, an orally active ATP-sensitive K+ channel (KATP) inhibitor used in diabetes and obesity research. It inhibits P-glycoprotein and directly binds and blocks SUR1 subunits of KATP. This compound interferes with mitochondrial bioenergetics and can induce autophagy. It is suitable as a tracer and internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits P-glycoprotein
  • Binds and blocks SUR1 subunits of KATP
  • Interferes with mitochondrial bioenergetics
  • Can induce autophagy