Learn More
Medchemexpress LLC Glyburide-d3 | 1219803-02-7 | C23H25D3ClN3O5S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15206S11MG
Encompass_Preferred
Glyburide-d3 is a deuterium-labeled version of Glibenclamide, an orally active inhibitor of ATP-sensitive K+ channels (KATP). It is used in research related to diabetes and obesity, inhibiting P-glycoprotein and blocking SUR1 subunits of KATP. Glyburide-d3 also impacts mitochondrial bioenergetics and can induce autophagy.
  • Deuterium labeled version of Glibenclamide.
  • Orally active ATP-sensitive K+ channel (KATP) inhibitor.
  • Used in research for diabetes and obesity.
  • Inhibits P-glycoprotein.
  • Blocks SUR1 subunits of KATP.
  • Inhibits cystic fibrosis transmembrane conductance regulator protein (CFTR).
  • Impacts mitochondrial bioenergetics.
  • Can induce autophagy.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-73677