Learn More
Cayman Chemical GlyburIde-d3 5mg
SDP
Catalog No. 502936482
Encompass_Preferred
An internal standard for the quantification of glyburide by GC- or LC-MS
Catalog No. 50-293-6482 Supplier Cayman Chemical Supplier No. 257405 MG
An internal standard for the quantification of glyburide by GC- or LC-MS