Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glyburide-d3 (deuterium-labeled glibenclamide) | 1219803-02-7 | MFCD06658648 | 99.6% | 497.02 | C23H25D3ClN3O5S | 10 MG
SDP

Catalog No. 5000285895
Encompass_Preferred

Glyburide-d3 is the deuterium-labeled isotopologue of glibenclamide supplied as a solid research reagent and analytical standard. It serves as a labeled internal standard and is used in studies of ATP-sensitive K+ (KATP) channel activity and related analytical workflows.

  • Deuterium-labeled internal standard for analytical assays.
  • High purity: 99.64%.
  • CAS number: 1219803-02-7.
  • Molecular weight: 497.02; chemical formula: C23H25D3ClN3O5S.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Solubility: DMSO 100 mg/mL; H2O 0.1 mg/mL (ultrasonic/warming recommended).

Catalog No. 50-002-85895 Supplier Medchemexpress LLC Supplier No. HY15206S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glyburide-d3 is the deuterium-labeled isotopologue of glibenclamide supplied as a solid research reagent and analytical standard. It serves as a labeled internal standard and is used in studies of ATP-sensitive K+ (KATP) channel activity and related analytical workflows.

  • Deuterium-labeled internal standard for analytical assays.
  • High purity: 99.64%.
  • CAS number: 1219803-02-7.
  • Molecular weight: 497.02; chemical formula: C23H25D3ClN3O5S.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Solubility: DMSO 100 mg/mL; H2O 0.1 mg/mL (ultrasonic/warming recommended).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.