Learn More
Selleck Chemical LLC Glyburide Glibenclamide-50mg
SDP
Catalog No. 502389586
Encompass_Preferred
Glyburide (Glibenclamide) is a known blocker of vascular ATP-sensitive K+ channels (KATP), used in the treatment of type 2 diabetes.
Catalog No. 50-238-9586 Supplier Selleck Chemical LLC Supplier No. S171650MG
Glyburide (Glibenclamide) is a known blocker of vascular ATP-sensitive K+ channels (KATP), used in the treatment of type 2 diabetes.