Learn More
AdipoGen Glyburide USP
SDP
Supplier:  AdipoGen AGCR13613G001
Encompass_Preferred
Chemical. CAS 10238-21-8. Formula C23H28ClN3O5S. MW 494. Antidiabetic compound. Binds to and activates the ATP-sensitive potassium channels KATP inhibitory regulatory subunit sulfonylurea receptor 1 SUR1. Causes cell membrane depolarization and opening of voltage-dependent calcium channel. Increases intracellular calcium and stimulates insulin secretion in beta cells NLRP3 inflammasome inhibitor. Broad-spectrum ATP-binding cassette ABC transporter inhibitor. Shown to have anti-leishmanial activity. Potential inhibitor of collagenases.
Catalog No. 50-256-2267