Learn More
Raybiotech Inc Glycan Array 100
Catalog No. 501950318
Encompass_Preferred
Detection of binding partners for 100 different glycans in purified proteins, serum, plasma, cell culture supernatants, cell/tissue lysates or other body fluids. View protein list on RayBiotech’s website – GA-Glycan-100-1
Catalog No. 50-195-0318 Supplier Raybiotech Inc Supplier No. GAGLYCAN1001
Detection of binding partners for 100 different glycans in purified proteins, serum, plasma, cell culture supernatants, cell/tissue lysates or other body fluids. View protein list on RayBiotech’s website – GA-Glycan-100-1