Learn More
Crystal Chem Inc Glycated Serum Proteins (GSP) Assay Kit, 96-tests
SDP
Catalog No. 501947666
Encompass_Preferred
The Glycated Serum Protein Assay is used to quantify levels of glycated serum protein in human serum in less than 10 min. Highlights: Provides short-term to medium-term (past 2-3 week period) average blood glucose levels. Complementary to HbA1c in diagnosis and screening of diabetes. Excellent correlation to ELISA method. Can be run manually or fully automated on a wide range of instruments. Less than 15 minute procedure. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7666 Supplier Crystal Chem Inc Supplier No. 80109
The Glycated Serum Protein Assay is used to quantify levels of glycated serum protein in human serum in less than 10 min. Highlights: Provides short-term to medium-term (past 2-3 week period) average blood glucose levels. Complementary to HbA1c in diagnosis and screening of diabetes. Excellent correlation to ELISA method. Can be run manually or fully automated on a wide range of instruments. Less than 15 minute procedure. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.