Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycerol, 56-81-5, MFCD00004722, 100 mL
Catalog No. NC0117244
Encompass
Linear Formula: HOCH2CH(OH)CH2OH, Molecular Weight: 92.09, 99%, BP: 182 °C/20 mmHg, MP: 20 °C, Synonym: 1,2,3-Propanetriol, Glycerin.
Catalog No. NC0117244 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G5516100ML
Linear Formula: HOCH2CH(OH)CH2OH, Molecular Weight: 92.09, 99%, BP: 182 °C/20 mmHg, MP: 20 °C, Synonym: 1,2,3-Propanetriol, Glycerin.