Learn More
U.S. Pharmacopeia Glycerin, 56-81-5, MFCD00004722, 2mL
Catalog No. NC9018607
Encompass
Molecular formula C3-H8-O3, Molecular weight 92.09 g/mol, Melting Point 62.6 - 64.4 °F (17 - 18 °C), Boiling Point 554 °F (290 °C) 101.325 kPa, Synonyms: Glycerolum, Glycerol, 1,2,3-Propanetriol
Catalog No. NC9018607 Supplier U.S. Pharmacopeia Supplier No. 1295607
Molecular formula C3-H8-O3, Molecular weight 92.09 g/mol, Melting Point 62.6 - 64.4 °F (17 - 18 °C), Boiling Point 554 °F (290 °C) 101.325 kPa, Synonyms: Glycerolum, Glycerol, 1,2,3-Propanetriol