Learn More
LAB ALLEY Lab Alley Glycerin 99.5% ACS Reagent Grade
SDP
Catalog No. NC2602857
Encompass
Glycerin is found in large amounts in vegetable and animal fats. It appears as a colorless odorless syrup-like liquid at room temperature. It is a nonvolatile liquid with a pleasant taste and easily soluble in alcohol and water. It is naturally hygroscopic with a high boiling point at ambient conditions. Reagent Grade chemicals are also known as the American Chemical Society (ACS) Grade chemicals. On the basis of its high purity our Glycerin ACS Reagent Grade 99.5 is a certified as the ACS Reagent Grade chemical. Lab Alley is selling its premium quality Glycerin ACS Reagent Grade 99.5% in the United States of America (USA) and highly recommends using it in food medicines and high-quality research labs.
Catalog No. NC2602857 Supplier LAB ALLEY Supplier No. GLA4L
Glycerin is found in large amounts in vegetable and animal fats. It appears as a colorless odorless syrup-like liquid at room temperature. It is a nonvolatile liquid with a pleasant taste and easily soluble in alcohol and water. It is naturally hygroscopic with a high boiling point at ambient conditions. Reagent Grade chemicals are also known as the American Chemical Society (ACS) Grade chemicals. On the basis of its high purity our Glycerin ACS Reagent Grade 99.5 is a certified as the ACS Reagent Grade chemical. Lab Alley is selling its premium quality Glycerin ACS Reagent Grade 99.5% in the United States of America (USA) and highly recommends using it in food medicines and high-quality research labs.