Learn More
Description
Glycerin USP is a clear, colorless, syrupy liquid. Doesn't have more than a slight characteristic odor, which is neither harsh nor disagreeable. It is hygroscopic.
Alternative name(s): Glycerol
Specifications
Specifications
| Chemical Name or Material | Glycerin USP |
| Packaging | Bottle, 500mL, Amber, PET, Boston Round, 28-400 |
| CAS | 56-81-5 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 92.11 |
| Quantity | 500 mL |
| Percent Purity | 99 to 101 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
