Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Glycerin USP
SDP

Catalog No. 14185559 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-559 4 L
14-185-270 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-185-559 Supplier Medisca Supplier No. 061301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycerin USP is a clear, colorless, syrupy liquid. Doesn't have more than a slight characteristic odor, which is neither harsh nor disagreeable.

Glycerin USP is a clear, colorless, syrupy liquid. Doesn't have more than a slight characteristic odor, which is neither harsh nor disagreeable. It is hygroscopic.

Alternative name(s): Glycerol

Specifications

Chemical Name or Material Glycerin USP
Packaging Jug, 4L, White, HDPE, Square, 38-400, TE Pano Neck
CAS 56-81-5
Type Drug Standard
Molecular Weight (g/mol) 92.11
Quantity 4 L
Percent Purity 99 to 101 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.