Learn More
Mitegen Glycerol (Glycerin), 100% Purity, 100 mL Bottle, For Cryoprotection, Solvent & General Lab Use
SDP
Catalog No. NC1381201
Encompass
This high-purity Glycerol, also known as Glycerin, is supplied as a 100% v/v solution in a convenient 100 mL bottle, making it an indispensable reagent for a wide array of laboratory applications. With a molecular weight of 92.09, this versatile compound (CAS: 56-81-5, MFCD: 00004722) is widely utilized across biochemistry, molecular biology, and crystallography. It is particularly effective as a cryoprotectant, helping to prevent ice crystal formation and protect delicate biological samples during freezing and storage at ultra-low temperatures. Beyond cryopreservation, Glycerol serves as a general solvent for various organic and inorganic substances, a humectant to maintain moisture, and a viscosity modifier in countless experimental protocols. Its high purity ensures reliable and reproducible results for sensitive assays and demanding research needs.
Catalog No. NC1381201 Supplier Mitegen Supplier No. MCSS188
This high-purity Glycerol, also known as Glycerin, is supplied as a 100% v/v solution in a convenient 100 mL bottle, making it an indispensable reagent for a wide array of laboratory applications. With a molecular weight of 92.09, this versatile compound (CAS: 56-81-5, MFCD: 00004722) is widely utilized across biochemistry, molecular biology, and crystallography. It is particularly effective as a cryoprotectant, helping to prevent ice crystal formation and protect delicate biological samples during freezing and storage at ultra-low temperatures. Beyond cryopreservation, Glycerol serves as a general solvent for various organic and inorganic substances, a humectant to maintain moisture, and a viscosity modifier in countless experimental protocols. Its high purity ensures reliable and reproducible results for sensitive assays and demanding research needs.