Learn More
Medchemexpress LLC Glycerol-13C-1 | 82425-96-5 | 99.3% | 50 MG
SDP
Catalog No. 5000413096
Encompass_Preferred
Glycerol-13C-1 (90% in water) is a 13C labeled Glycerol. Glycerol is utilized in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is intended for research use only and not sold to patients.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.
Catalog No. 50-004-13096 Supplier Medchemexpress LLC Supplier No. HYB1659S250MG
Glycerol-13C-1 (90% in water) is a 13C labeled Glycerol. Glycerol is utilized in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is intended for research use only and not sold to patients.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.