Learn More
Medchemexpress LLC Glycerol-13C2 | 102088-01-7 | 99% | 100 MG
SDP
Catalog No. 5000412725
Encompass_Preferred
Glycerol-13C2 is the 13C labeled Glycerol, used in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is for research use only.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes like 13C incorporated into drug molecules can be used as tracers for quantitation during drug development
Catalog No. 50-004-12725 Supplier Medchemexpress LLC Supplier No. HYB1659S4100MG
Glycerol-13C2 is the 13C labeled Glycerol, used in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is for research use only.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes like 13C incorporated into drug molecules can be used as tracers for quantitation during drug development