Learn More
Bioworld Glycerol (RNAse, DNAse and Protease Free), 100 mL
Catalog No. 50256031
Encompass_Preferred
GlycerolGlycerol(orglycerine,glycerin) is a polyolcompound. It is colorless, odorless,viscousliquid that is widely used inbiological applications.Synonym: glycerinCommon reagent used in tissue culture media for bacteria and enzyme storage.Useful for mammalian cell transfection and polyacrylamide gel preservation protocolsUseful for sample preparation/protein stabilization and as a storage buffer componentSpecifications:USP GradeSpecific gravity: 1.249Heavy metals (as Pb) <0.0005%RNAse, Protease Activity: None detectedNon-sterileIUPAC_Name: Propan-1,2,3-triolPH Value: 5.5 - 8RTECS Number: MA8050000Viscosity: 1.2 Pas
Catalog No. 50-256-031 Supplier Bioworld Supplier No. 407000403
Add to Cart
Add to Cart
GlycerolGlycerol(orglycerine,glycerin) is a polyolcompound. It is colorless, odorless,viscousliquid that is widely used inbiological applications.Synonym: glycerinCommon reagent used in tissue culture media for bacteria and enzyme storage.Useful for mammalian cell transfection and polyacrylamide gel preservation protocolsUseful for sample preparation/protein stabilization and as a storage buffer componentSpecifications:USP GradeSpecific gravity: 1.249Heavy metals (as Pb) <0.0005%RNAse, Protease Activity: None detectedNon-sterileIUPAC_Name: Propan-1,2,3-triolPH Value: 5.5 - 8RTECS Number: MA8050000Viscosity: 1.2 Pas