Learn More
Medchemexpress LLC Glycerol-3-phosphate dehydrogenase, rabbit muscle | 9075-65-4 | 100-300 U/mg protein | 35,780 Da | 1 KU
SDP
Catalog No. 5000400337
Encompass_Preferred
Glycerol-3-phosphate dehydrogenase, rabbit muscle (GPDH) is an enzyme that catalyzes the reversible redox conversion of dihydroxyacetone phosphate to sn-glycerol 3-phosphate. It functions as a glycolytic enzyme and is crucial for the re-oxidation of NADH to NAD+ and for energy supply. The product appears as a liquid, colorless to light yellow. It is intended for research use only.
  • Catalyzes reversible redox conversion of dihydroxyacetone phosphate
  • Crucial for re-oxidation of NADH to NAD+
  • Important for energy supply
  • Liquid, colorless to light yellow appearance
  • For research use only
Catalog No. 50-004-00337 Supplier Medchemexpress LLC Supplier No. HYP27651KU
Glycerol-3-phosphate dehydrogenase, rabbit muscle (GPDH) is an enzyme that catalyzes the reversible redox conversion of dihydroxyacetone phosphate to sn-glycerol 3-phosphate. It functions as a glycolytic enzyme and is crucial for the re-oxidation of NADH to NAD+ and for energy supply. The product appears as a liquid, colorless to light yellow. It is intended for research use only.
  • Catalyzes reversible redox conversion of dihydroxyacetone phosphate
  • Crucial for re-oxidation of NADH to NAD+
  • Important for energy supply
  • Liquid, colorless to light yellow appearance
  • For research use only