Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol-3-phosphate oxidase, MP Biomedicals™

Catalog No. ICN15724090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 U
50 U
500 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15724090 500 U
ICN15724050 50 U
ICN15724080 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15724090 Supplier MP Biomedicals, Inc Supplier No. 0215724090
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9046-28-0
Synonym sn-Glycerol-3-phosphate, oxygen 2-oxidoreductase

Specifications

Color Yellow
Quantity 500 U
Physical Form Lyophilized Powder
Chemical Name or Material Glycerol-3-phosphate oxidase
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.