Learn More
Medchemexpress LLC Glycerol | 56-81-5 | 98.9% | 250 MG
SDP
Catalog No. 5000414254
Encompass_Preferred
Glycerol (Standard) is the analytical standard of Glycerol, intended for research and analytical applications. It is used in sample preparation and gel formation for polyacrylamide gel electrophoresis, and is suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Used as an analytical standard
  • Suitable for research and analytical applications
  • Supports sample preparation and gel formation
  • Compatible with HPLC, GC, and MS experiments
Catalog No. 50-004-14254 Supplier Medchemexpress LLC Supplier No. 250MGHYB1659R
Glycerol (Standard) is the analytical standard of Glycerol, intended for research and analytical applications. It is used in sample preparation and gel formation for polyacrylamide gel electrophoresis, and is suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Used as an analytical standard
  • Suitable for research and analytical applications
  • Supports sample preparation and gel formation
  • Compatible with HPLC, GC, and MS experiments