Learn More
Medchemexpress LLC Glycerol | 56-81-5 | 99.9% | 500 ML
SDP
Catalog No. 5000413171
Encompass_Preferred
Glycerol is a versatile chemical used in various laboratory applications. It is commonly employed in sample preparation and gel formation for polyacrylamide gel electrophoresis, helping to prevent the dissociation of nucleosomes and other protein-DNA complexes during the process. This product is suitable for research use only.
  • Used in polyacrylamide gel electrophoresis
  • Helps prevent dissociation of nucleosomes and protein-DNA complexes
  • Induces acute renal failure in rat models for research purposes
Catalog No. 50-004-13171 Supplier Medchemexpress LLC Supplier No. HYB1659500ML
Glycerol is a versatile chemical used in various laboratory applications. It is commonly employed in sample preparation and gel formation for polyacrylamide gel electrophoresis, helping to prevent the dissociation of nucleosomes and other protein-DNA complexes during the process. This product is suitable for research use only.
  • Used in polyacrylamide gel electrophoresis
  • Helps prevent dissociation of nucleosomes and protein-DNA complexes
  • Induces acute renal failure in rat models for research purposes