Learn More
Randox GLYCEROL 6X15ML
SDP
Catalog No. NC2734236
Encompass_Preferred
GLYCEROL 6x15ML; GPO-PAP method; Lyophilised reagents; Measuring range 14.5 – 2545 umol/l
Catalog No. NC2734236 Supplier Randox Supplier No. GY105
GLYCEROL 6x15ML; GPO-PAP method; Lyophilised reagents; Measuring range 14.5 – 2545 umol/l