Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycerol, 56-81-5, MFCD00004722, 100 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G9012100ML
Encompass
Linear Formula: HOCH2CH(OH)CH2OH, Molecular Weight: 92.09, ≥99.5%, Synonym: 1,2,3-Propanetriol, Glycerin. Glycerol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel’s sample well. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component.
Catalog No. NC0718487