Learn More
Research Products International Corp Glycerol ACS 10 L
SDP
Catalog No. 502114043
Encompass_Preferred
Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Also used in casting gradient gels, as a protein stabilizer and storage buffer component.
Catalog No. 50-211-4043 Supplier Research Products International Corp Supplier No. G2202510.0
Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Also used in casting gradient gels, as a protein stabilizer and storage buffer component.