Learn More
Research Products International Corp Glycerol ACS 20 L
SDP
Catalog No. 501977103
Encompass_Preferred
Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Also used in casting gradient gels, as a protein stabilizer and storage buffer component.
Catalog No. 50-197-7103 Supplier Research Products International Corp Supplier No. G2202520.0
Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Also used in casting gradient gels, as a protein stabilizer and storage buffer component.