Learn More
Research Products International Corp Glycerol, ACS Grade, 4 Liters
SDP
Supplier:  Research Products International Corp G220254.0
Encompass_Preferred
Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Also used in casting gradient gels, as a protein stabilizer and storage buffer component.
Catalog No. 50-712-743