Learn More
Research Products International Corp Glycerol, ACS Grade, 500 Milliliters
SDP
Supplier:  Research Products International Corp G220250.5
Encompass_Preferred
Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Also used in casting gradient gels, as a protein stabilizer and storage buffer component.
Catalog No. 50-712-742