Learn More
Roche Diagnostics Glycerol Bio, 200 tests
Supplier:  Roche Diagnostics 07374607001
Encompass
Glycerol can be used as source of carbon and energy in microbial fermentation. In order to provide sufficient supply and to avoid inhibition by high concentrations, the optimal feeding with glycerol should be according to the actual consumption. For control of culture media and for monitoring in a culture process, the glycerol concentration can be determined on a Cedex Bio or Cedex Bio HT Analyzer with high reliability and convenience.
Catalog No. NC1351986