Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycerol BioXtra 99 GC
Catalog No. 502906983
Encompass
Glycerol is odourless colorless viscous in nature and exists as a sweet tasting liquid. It can be derived naturally as well as from petrochemical feedstock. Glycerol has a wide variety of applications and is one of the most valuable and versatile chemical substances in nature. It can be used as an emollient solvent sweetening agent in pharmaceutical formulations cosmetics foodstuffs and toiletries. It is very stable and can be easily stored under normal temperature also it is non-irritating and has no adverse impact to the environment.
Catalog No. 50-290-6983 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G62794X4L
Glycerol is odourless colorless viscous in nature and exists as a sweet tasting liquid. It can be derived naturally as well as from petrochemical feedstock. Glycerol has a wide variety of applications and is one of the most valuable and versatile chemical substances in nature. It can be used as an emollient solvent sweetening agent in pharmaceutical formulations cosmetics foodstuffs and toiletries. It is very stable and can be easily stored under normal temperature also it is non-irritating and has no adverse impact to the environment.