Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bioworld GLYCEROL BUF(50%) PBS, 125ML

Catalog No. NC2001895
Encompass_Preferred

Glycerol Buffer (50%) in PBS, pH 7.4, Sterile. 125ml BIOWORLD

Catalog No. NC2001895 Supplier Bioworld Supplier No. 401203561
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Glycerol Buffer (50%) in PBS, pH 7.4, Sterile. 125ml BIOWORLD

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.