Learn More
Cayman Chemical Glycerol Cell-Based Assay Kit, 1 ea
SDP
Catalog No. 502886977
Encompass_Preferred
Cayman’s Glycerol Cell-Based Assay provides a convenient tool for studying triglycerides/fatty acid cycling and its regulation in adipocytes or hepatocytes. This kit will allow investigators to screen compounds involved in lipid storage and metabolism. Chloroquine is included in the kit as a positive control for screening pharmaceuticals that induce lipid droplet accumulation and free glycerol release from hepatocytes.
Catalog No. 50-288-6977 Supplier Cayman Chemical Supplier No. 100117251 EA
Cayman’s Glycerol Cell-Based Assay provides a convenient tool for studying triglycerides/fatty acid cycling and its regulation in adipocytes or hepatocytes. This kit will allow investigators to screen compounds involved in lipid storage and metabolism. Chloroquine is included in the kit as a positive control for screening pharmaceuticals that induce lipid droplet accumulation and free glycerol release from hepatocytes.