Learn More
Medchemexpress LLC Glycerol-d3 | 7325-16-8 | 1 G
SDP
Catalog No. 5000413281
Encompass_Preferred
Glycerol-d3 is the deuterium labeled Glycerol, utilized in sample preparation and gel formation for polyacrylamide gel electrophoresis. Stable heavy isotopes, like deuterium, are incorporated into drug molecules as tracers for quantitation during drug development.
  • Can serve as a tracer
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Deuteration may influence the pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-004-13281 Supplier Medchemexpress LLC Supplier No. HYB1659S81G
Add to Cart
Add to Cart
Glycerol-d3 is the deuterium labeled Glycerol, utilized in sample preparation and gel formation for polyacrylamide gel electrophoresis. Stable heavy isotopes, like deuterium, are incorporated into drug molecules as tracers for quantitation during drug development.
  • Can serve as a tracer
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Deuteration may influence the pharmacokinetic and metabolic profiles of drugs
  • For research use only