Learn More
Medchemexpress LLC Glycerol-d8 (1,2,3-propanetriol-d8) | 7325-17-9 | MFCD00084149 | 98.3% | 100.14 | C3D8O3 | 50 MG
SDP
Catalog No. 5000282601
Encompass_Preferred
Glycerol-d8 (1,2,3-Propanetriol-d8) is the deuterium-labeled analogue of glycerol used as a stable isotope tracer and internal standard in analytical and research workflows. It is supplied at high chemical and isotopic purity for use in NMR spectroscopy, mass spectrometry, and metabolic tracing. This product is intended for research use only.
  • High isotopic labeling for clear mass shifts and NMR signals.
  • High chemical purity to minimize background interference.
  • Compatible with NMR and mass spectrometry analytical workflows.
  • Useful as an internal standard for accurate quantitation.
  • Stable liquid form for easy sample preparation and handling.
Catalog No. 50-002-82601 Supplier Medchemexpress LLC Supplier No. HYW12757250MG
Glycerol-d8 (1,2,3-Propanetriol-d8) is the deuterium-labeled analogue of glycerol used as a stable isotope tracer and internal standard in analytical and research workflows. It is supplied at high chemical and isotopic purity for use in NMR spectroscopy, mass spectrometry, and metabolic tracing. This product is intended for research use only.
  • High isotopic labeling for clear mass shifts and NMR signals.
  • High chemical purity to minimize background interference.
  • Compatible with NMR and mass spectrometry analytical workflows.
  • Useful as an internal standard for accurate quantitation.
  • Stable liquid form for easy sample preparation and handling.