Learn More
Medchemexpress LLC Glycerol dehydrogenase | 9028-14-2 | 500 U
SDP
Catalog No. 5000400409
Encompass_Preferred
Glycerol dehydrogenase is a polyol dehydrogenase that catalyzes the NAD-dependent oxidation of glycerol to dihydroxyacetone. It can be utilized for glycerol metabolism in various microorganisms.
  • Catalyzes NAD-dependent oxidation of glycerol to dihydroxyacetone.
  • Utilized for glycerol metabolism in microorganisms.
  • Appearance: solid.
  • Color: white to off-white.
  • For research use only.
Catalog No. 50-004-00409 Supplier Medchemexpress LLC Supplier No. HYP2725500U
Glycerol dehydrogenase is a polyol dehydrogenase that catalyzes the NAD-dependent oxidation of glycerol to dihydroxyacetone. It can be utilized for glycerol metabolism in various microorganisms.
  • Catalyzes NAD-dependent oxidation of glycerol to dihydroxyacetone.
  • Utilized for glycerol metabolism in microorganisms.
  • Appearance: solid.
  • Color: white to off-white.
  • For research use only.