Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycerol diglycidyl ether

Catalog No. 475734500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-573-4500ML 500 mL
47-573-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-573-4500ML Supplier Sigma Aldrich Supplier No. 475734500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycerol diglycidyl ether (GDE) is an aliphatic epoxy monomer that can be used as a diepoxy crosslinker. It has a low viscous epoxy matrix that can be cured with aliphatic amines to form a thermosetting material with good flexibility. Its properties include low shrinkage, good adhesion, and good thermo-mechanical properties.GDE can be used in the formation of epoxy materials, which can further be used in electrical instruments and biodegradable plastics.

Specifications

CAS 27043-36-3
Density 1.229 g/mL (at 25°C)
Molecular Formula C9H16O5
Refractive Index n20/D 1.482 (literature)
Linear Formula C9H16O5
Quantity 500 mL
Synonym Glycerin diglycidyl ether; Glycerol diglycidyl ether
Molecular Weight (g/mol) 204.22
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.