Learn More
Cayman Chemical Glycerol Enzyme MIxture 1ea
SDP
Catalog No. 502942232
Encompass_Preferred
Glycerol enzyme mixture has been tested and formulated to work exclusively with Cayman’s Glycerol Colorimetric Assay Kit (Item No. 10010755).
Catalog No. 50-294-2232 Supplier Cayman Chemical Supplier No. 4007891 EA
Add to Cart
Add to Cart
Glycerol enzyme mixture has been tested and formulated to work exclusively with Cayman’s Glycerol Colorimetric Assay Kit (Item No. 10010755).