Learn More
Electron Microscopy Sciences EMS Glycerol Fluoro Mount w/Para Phenylenediamine Anti-fading 20 ML
SDP
Catalog No. 501927434
Encompass_Preferred
EMS Glycerol Fluorescence Mount with strong anti-fading agent, 1, 4-phenylenediamine (PPD) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 degree C in the dark.
This medium contains phenylenediamine and is not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PE), phyocyanin (PC), and allophycocyanin (APC).
Use for immunofluorescence and confocal microscopy.
The ready to use mounting medium has a dark coffee color that does not interfere with Immunofluorescence.
Refractive Index is 1.4563. Storage at 2-8 degree C is recommended. Protect from light, DO NOT FREEZE.
Catalog No. 50-192-7434 Supplier Electron Microscopy Sciences Supplier No. 1798970
EMS Glycerol Fluorescence Mount with strong anti-fading agent, 1, 4-phenylenediamine (PPD) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 degree C in the dark.
This medium contains phenylenediamine and is not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PE), phyocyanin (PC), and allophycocyanin (APC).
Use for immunofluorescence and confocal microscopy.
The ready to use mounting medium has a dark coffee color that does not interfere with Immunofluorescence.
Refractive Index is 1.4563. Storage at 2-8 degree C is recommended. Protect from light, DO NOT FREEZE.