Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycerol formal

Catalog No. 49920100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-920-100ML 100 mL
49-920-500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 49-920-100ML Supplier Sigma Aldrich Supplier No. 49920100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Glycerol formal (gf) is a light glycerol acetal that is composed of ~60% 5-hydroxy-1,3-dioxane and ~40% 4-hydroxymethyl-1,3-dioxolane
  • It can be prepared by reacting glycerol and formaldehyde in the presence of acid catalyst
  • The potential of gf as an injection solvent for use in toxicity testing has been reported.glycerol formal may be used in the multi-step synthesis of 9-[(2-hydroxy-1-(hydroxymethyl)ethoxy)methyl]guanine (2α-nor-2α-deoxyguanosine), an analog of acyclovir
  • It may be used in the preparation of 4-substituted dioxolanes

Specifications

CAS 99569-11-6
Density 1.203 g/mL (at 25°C (literature))
Boiling Point 192°C to 193°C (lit.)
Molecular Formula C4H8O3
Refractive Index n20/D 1.451 (literature); n20/D 1.451
Linear Formula C4H8O3
MDL Number MFCD10039829
Quantity 100 mL
Molecular Weight (g/mol) 104.1
Percent Purity ≥98.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.