Learn More
Meridian Life Science Glycerol-Free Taq HS High, 20ML
SDP
Catalog No. 5000856469
Encompass_Preferred
Supplied as a blend of high‑performance glycerol‑free Taq DNA Polymerase with a Taq hot-start antibody and is suitable for difficult, multiplex PCR that require fast amplification, whilst maintaining both high specificity and sensitivity.Glycerol-Free Taq HS High Conc. is ideal for PCR kit manufactures and PCR molecular test facilities, requiring large amounts of DNA polymerases. The highly concentrated Glycerol-Free Taq HS High Conc. allows for concentrated bulk production of PCR master mixes and the glycerol free format is useful when using automated pipetting robots and other applications where accurate pipetting of small volumes is crucial.
Catalog No. 50-008-56469 Supplier Meridian Life Science Supplier No. MDX170002
Supplied as a blend of high‑performance glycerol‑free Taq DNA Polymerase with a Taq hot-start antibody and is suitable for difficult, multiplex PCR that require fast amplification, whilst maintaining both high specificity and sensitivity.Glycerol-Free Taq HS High Conc. is ideal for PCR kit manufactures and PCR molecular test facilities, requiring large amounts of DNA polymerases. The highly concentrated Glycerol-Free Taq HS High Conc. allows for concentrated bulk production of PCR master mixes and the glycerol free format is useful when using automated pipetting robots and other applications where accurate pipetting of small volumes is crucial.