Learn More
Crescent Chemical Co Inc GLYCEROL FROM PLANT 87%
SDP
Catalog No. 50248478
Encompass_Preferred
Glycerol from plant 87 % molecular biology grade DNase/RNase not detected. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 39788.01 CAS 56-81-5
Catalog No. 50-248-478 Supplier Crescent Chemical Co Inc Supplier No. SE39788.01
Glycerol from plant 87 % molecular biology grade DNase/RNase not detected. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 39788.01 CAS 56-81-5