Learn More
Alkali Scientific Glycerol Gel Loading Dye, 5X (Contains 30% Glycerol), 5 Milliliters
SDP
Catalog No. 5000661773
Encompass_Preferred
Glycerol Gel Loading Dye, 5X (Contains 30% Glycerol), is an essential reagent for DNA sample loading in electrophoresis. Supplied in a 5mL vial, this dye offers the necessary viscosity and color to prepare DNA samples for gel electrophoresis. The glycerol component ensures that samples settle properly in the gel wells, while the color serves as an indicator for sample tracking during electrophoresis. The 5X concentration allows for easy dilution with the DNA samples, providing optimal results for DNA separation and visualization. This dye is commonly used in molecular biology laboratories for PCR amplification, sequencing, and other DNA-based assays.
Catalog No. 50-006-61773 Supplier Alkali Scientific Supplier No. YCE002CP
Glycerol Gel Loading Dye, 5X (Contains 30% Glycerol), is an essential reagent for DNA sample loading in electrophoresis. Supplied in a 5mL vial, this dye offers the necessary viscosity and color to prepare DNA samples for gel electrophoresis. The glycerol component ensures that samples settle properly in the gel wells, while the color serves as an indicator for sample tracking during electrophoresis. The 5X concentration allows for easy dilution with the DNA samples, providing optimal results for DNA separation and visualization. This dye is commonly used in molecular biology laboratories for PCR amplification, sequencing, and other DNA-based assays.