Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycerol Gelatin aqueous s
Catalog No. 502900114
Encompass
Glycerol alters the hygroscopic property of gelatin and its permeability to water vapour. Glycerol Gelatin or glycerine jelly medium is an aqueous slide mounting media used for mounting samples in microscopy and histology.
Catalog No. 50-290-0114 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GG115ML
Glycerol alters the hygroscopic property of gelatin and its permeability to water vapour. Glycerol Gelatin or glycerine jelly medium is an aqueous slide mounting media used for mounting samples in microscopy and histology.