Learn More
Glycerol Gelatin, Sigma-Aldrich
Catalog No. 01258932 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
15 mL
Catalog No. Volume (Metric)
01-258-932 15 mL
1 options
Catalog No. 01-258-932 Supplier MilliporeSigma Supelco Supplier No. GG110X15ML
Add to Cart
Add to Cart
Glycerol alters the hygroscopic property of gelatin and its permeability to water vapour. Glycerol Gelatin or glycerine jelly medium is an aqueous slide mounting media, used for mounting samples in microscopy and histology.
10 x 15 mL per pack
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 15 mL