Learn More
Electron Microscopy Sciences EMS Glycerol Mounting Medium w/ DAPI & DABCO 20 ML
SDP
Catalog No. 501927432
Encompass_Preferred
EMS Glycerol with DAPI and DABCO is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. It prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Fluorescence is retained during prolonged storage at 4 deg C in the dark.
This mounting medium is fortified with DAPI which is a counter-stain for DNA. This product is to be used in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360nm and emits at 460nm, producing a blue fluorescence. RNA is also stained with DAPI.
Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC).
Reagent: Ready to use mounting medium with anti-fading agent
Refractive Index: 1.4617
Catalog No. 50-192-7432 Supplier Electron Microscopy Sciences Supplier No. 1798960
EMS Glycerol with DAPI and DABCO is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. It prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Fluorescence is retained during prolonged storage at 4 deg C in the dark.
This mounting medium is fortified with DAPI which is a counter-stain for DNA. This product is to be used in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360nm and emits at 460nm, producing a blue fluorescence. RNA is also stained with DAPI.
Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC).
Reagent: Ready to use mounting medium with anti-fading agent
Refractive Index: 1.4617