Learn More
Electron Microscopy Sciences EMS Glycerol Mounting Medium 100 ML
SDP
Supplier:  Electron Microscopy Sciences 1798940
Encompass_Preferred
This aqueous mounting medium made with glycerol is for preserving fluorescence of tissue and cell smears. It is excellent for preserving thick sections that can not be mounted with mounting medium or fresh frozen tissues containing lipids, e.g. brain tissue.
This mounting medium is also useful for immunofluorescence. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy dyes, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine and Redox. The fluorescence is retained during prolonged storage at 4 degree C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes.
Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC).
Reagent: Ready to use mounting medium for IHC and immunofluorescence (IF).
Refractive Index: 1.472. Storage at temperature of 2-8 degree C is recommended. Protect from light.
Catalog No. 50-192-7428