Learn More
Electron Microscopy Sciences EMS Glycerol Mounting Medium w/ DABCO 30 ML
SDP
Catalog No. 501927430
Encompass_Preferred
EMS Glycerol with anti-fading agent 1, 4-Diazobicyclo-(2,2,2-octane (DABCO) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Fluorescence is retained during prolonged storage at 4 degree C in the dark. Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC).
Application: Immunofluorescence, confocal microscopy.
Reagent: Ready to use mounting medium with anti-fading agent.
Catalog No. 50-192-7430 Supplier Electron Microscopy Sciences Supplier No. 1798950
EMS Glycerol with anti-fading agent 1, 4-Diazobicyclo-(2,2,2-octane (DABCO) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Fluorescence is retained during prolonged storage at 4 degree C in the dark. Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC).
Application: Immunofluorescence, confocal microscopy.
Reagent: Ready to use mounting medium with anti-fading agent.