Learn More
Polysciences, Inc. Glycerol Monomethacrylate, 25g
SDP
Catalog No. NC1624929
Encompass
Glycerol methacrylate (GMA) is a hydrophilic monomer useful in hydrogel preparation. Hydrogels, hydrophilic polymer networks, have been extensivelystudied becauseof their potential in biomedical, industrial and environmental applications. 5919-74-4, 25g
Catalog No. NC1624929 Supplier Polysciences, Inc. Supplier No. 0418025
Glycerol methacrylate (GMA) is a hydrophilic monomer useful in hydrogel preparation. Hydrogels, hydrophilic polymer networks, have been extensivelystudied becauseof their potential in biomedical, industrial and environmental applications. 5919-74-4, 25g