Learn More
TARGETMOL CHEMICALS INC Glycerol phenylbutyrate 100MG
SDP
Catalog No. 502876253
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Glycerol phenylbutyrate (HPN-100) (GPB) is a new generation ammonia scavenger drug and it also is a sigma-2 (σ2) receptor ligand (pKi 8.02). Purity 99.74%
Catalog No. 50-287-6253 Supplier TARGETMOL CHEMICALS INC Supplier No. T15393100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Glycerol phenylbutyrate (HPN-100) (GPB) is a new generation ammonia scavenger drug and it also is a sigma-2 (σ2) receptor ligand (pKi 8.02). Purity 99.74%